Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHB
Entrez Gene ID
818036
Gene Name
Homeobox-leucine zipper family protein / lipid-binding START domain-containing protein
Gene Aliases
ARABIDOPSIS THALIANA HOMEOBOX PROTEIN 14, AT2G34710, ATHB-14, ATHB14, PHABULOSA, PHABULOSA 1D, PHB-1D, T29F13.8, T29F13_8
Chromosome Location
Chr.2: 14639323 - 14644355 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129025.4 NP_181018.1 15 - 16 2630 62
GenBank mRNA AJ441291.1 - 15 - 16 2268 62
AY099791.1 - 15 - 16 2546 62
BT000335.1 - 15 - 16 2268 62

View Full Product Details