Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TO1
Entrez Gene ID
818065
Gene Name
thioredoxin O1
Gene Aliases
AT2G35010, ATO1, F19I3.24, F19I3_24, thioredoxin O1
Chromosome Location
Chr.2: 14754234 - 14756323 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084537.1 NP_001078006.1 4 - 5 567 131
NM_129053.6 NP_181046.1 4 - 5 604 131
GenBank mRNA BT025773.1 - 4 - 5 440 131

View Full Product Details