Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NPSN11
- Entrez Gene ID
-
818086
- Gene Name
- Putative plant snare 11
- Gene Aliases
-
AT2G35190, ATNPSN11, NSPN11, T4C15.14, T4C15_14, novel plant snare 11
- Chromosome Location
-
Chr.2: 14830626 - 14833417 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129072.4 |
NP_565800.1 |
4 - 5 |
861 |
96 |
View Full Product Details