Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EYA
Entrez Gene ID
818099
Gene Name
eyes absent-like protein
Gene Aliases
AT2G35320, ATEYA, EYES ABSENT homolog, T4C15.1, T4C15_1
Chromosome Location
Chr.2: 14866832 - 14868881 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129084.3 NP_565803.1 4 - 5 631 57
GenBank mRNA AF410332.1 - 4 - 5 560 57
AY054555.1 - 4 - 5 585 57
AY064618.1 - 3 - 4 499 57
AY149930.1 - 3 - 4 499 57

View Full Product Details