Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE29
Entrez Gene ID
818101
Gene Name
helicase domain-containing protein
Gene Aliases
AT2G35340, T32F12.28, T32F12_28, maternal effect embryo arrest 29
Chromosome Location
Chr.2: 14872265 - 14879739 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336552.1 NP_001323587.1 23 - 24 2695 68
NM_129086.4 NP_181077.3 27 - 28 3255 68

View Full Product Details