Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMA5
Entrez Gene ID
842650
Gene Name
heavy metal atpase 5
Gene Aliases
AT1G63440, F2K11.18, F2K11_18, heavy metal atpase 5
Chromosome Location
Chr.1: 23527570 - 23531226 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105023.2 NP_176533.1 5 - 6 2652 78

View Full Product Details