Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- mtACP2
- Entrez Gene ID
-
842836
- Gene Name
- mitochondrial acyl carrier protein 2
- Gene Aliases
-
AT1G65290, T8F5.6, T8F5_6, mitochondrial acyl carrier protein 2
- Chromosome Location
-
Chr.1: 24248727 - 24250436 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105202.5 |
NP_176708.1 |
1 - 2 |
277 |
100 |
View Full Product Details