Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSAL
Entrez Gene ID
826892
Gene Name
photosystem I subunit l
Gene Aliases
AT4G12800, PSI-L, T20K18.150, T20K18_150, photosystem I subunit l
Chromosome Location
Chr.4: 7521257 - 7522761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340808.1 NP_001328992.1 1 - 2 378 66
NM_117349.3 NP_193016.1 1 - 2 313 66
GenBank mRNA AF428447.1 - 1 - 2 202 66
AF446350.1 - 1 - 2 202 66
AY059949.1 - 1 - 2 208 66
AY087339.1 - 1 - 2 314 66
AY097427.1 - 1 - 2 166 66

View Full Product Details