Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MHK
Entrez Gene ID
826915
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT4G13020, F25G13.110, F25G13_110, PROTEIN KINASE
Chromosome Location
Chr.4: 7602989 - 7607155 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036545.1 NP_001031622.1 9 - 10 999 70
NM_001125502.1 NP_001118974.1 11 - 12 1283 70
NM_001203783.2 NP_001190712.1 11 - 12 1320 70
NM_001340823.1 NP_001329480.1 11 - 12 1293 70
NM_001340824.1 NP_001329479.1 11 - 12 1293 70
NM_001340825.1 NP_001329481.1 11 - 12 1320 70
NM_117371.1 NP_193038.1 9 - 10 999 70
NM_179039.4 NP_849370.1 11 - 12 1405 70
GenBank mRNA AF360324.1 - 11 - 12 1254 70
AY056332.1 - 9 - 10 786 70

View Full Product Details