Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB79
Entrez Gene ID
826980
Gene Name
myb domain protein 79
Gene Aliases
AT4G13480, AtMYB79, T6G15.30, T6G15_30, myb domain protein 79
Chromosome Location
Chr.4: 7836644 - 7837763 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117422.3 NP_193084.1 1 - 2 142 62
GenBank mRNA AY133705.1 - 1 - 2 111 62

View Full Product Details