Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XTH15
Entrez Gene ID
827051
Gene Name
xyloglucan endotransglucosylase/hydrolase 15
Gene Aliases
AT4G14130, DL3105C, FCAALL.173, XTR7, xyloglucan endotransglucosylase/hydrolase 15, xyloglucan endotransglycosylase 7
Chromosome Location
Chr.4: 8137020 - 8138392 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117490.4 NP_193149.2 1 - 2 465 75
GenBank mRNA AY045865.1 - 1 - 2 354 75
AY087282.1 - 1 - 2 354 75
BT020422.1 - 1 - 2 269 75
U43489.1 - 1 - 2 269 75

View Full Product Details