Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP97B3
Entrez Gene ID
827177
Gene Name
cytochrome P450, family 97, subfamily B, polypeptide 3
Gene Aliases
''cytochrome P450, AT4G15110, DL3600C, FCAALL.208, cytochrome P450, family 97, polypeptide 3, polypeptide 3'', subfamily B
Chromosome Location
Chr.4: 8629647 - 8633118 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117600.6 NP_193247.2 7 - 8 1106 66
GenBank mRNA AY062675.1 - 7 - 8 1014 66
BT002582.1 - 7 - 8 981 66

View Full Product Details