Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSLB06
Entrez Gene ID
827198
Gene Name
cellulose synthase-like B6
Gene Aliases
AT4G15320, ATCSLB06, ATCSLB6, CELLULOSE SYNTHASE LIKE B6, DL3705C, FCAALL.268, cellulose synthase-like B6
Chromosome Location
Chr.4: 8742639 - 8748025 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341028.1 NP_001328692.1 10 - 11 2080 153
NM_117620.1 NP_193267.1 10 - 11 2209 153

View Full Product Details