Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CLA1
- Entrez Gene ID
-
827230
- Gene Name
- Deoxyxylulose-5-phosphate synthase
- Gene Aliases
-
1-DEOXY-D-XYLULOSE 5-PHOSPHATE SYNTHASE, 1-DEOXY-D-XYLULOSE 5-PHOSPHATE SYNTHASE 2, 1-deoxy-D-xylulose 5-phosphate (DXP) synthase 1, AT4G15560, AtCLA1, CLA, CLOROPLASTOS ALTERADOS 1, DEF, DL3821W, DXPS2, DXS, DXS1, FCAALL.24
- Chromosome Location
-
Chr.4: 8883825 - 8887667 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117647.3 |
NP_193291.1 |
4 - 5 |
1257 |
67 |
GenBank mRNA |
BT002340.1 |
- |
4 - 5 |
864 |
67 |
View Full Product Details