Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP20
Entrez Gene ID
827513
Gene Name
ubiquitin-specific protease 20
Gene Aliases
AT4G17895, UBIQUITIN-SPECIFIC PROTEASE 20, ubiquitin-specific protease 20
Chromosome Location
Chr.4: 9939666 - 9942988 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117899.3 NP_567544.1 2 - 3 883 102
GenBank mRNA AF302668.1 - 2 - 3 854 102

View Full Product Details