Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IP5PII
- Entrez Gene ID
-
827526
- Gene Name
- myo-inositol polyphosphate 5-phosphatase 2
- Gene Aliases
-
4, 5)P3 5-PHOSPHATASE II, 5PTASE2, AT4G18010, AT5PTASE2, INOSITOL POLYPHOSPHATE 5-PHOSPHATASE II, INOSITOL(1, T6K21.190, T6K21_190, myo-inositol polyphosphate 5-phosphatase 2
- Chromosome Location
-
Chr.4: 9990988 - 9994524 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117911.4 |
NP_567547.1 |
9 - 10 |
2168 |
117 |
View Full Product Details