Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RFC1
Entrez Gene ID
832261
Gene Name
replication factor C1
Gene Aliases
AT5G22010, AtRFC1, replication factor C 1, replication factor C1
Chromosome Location
Chr.5: 7280321 - 7287434 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343715.1 NP_001330213.1 5 - 6 860 92
NM_147883.6 NP_680188.1 5 - 6 860 92
GenBank mRNA BT014965.1 - 3 - 4 658 92

View Full Product Details