Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPB2
Entrez Gene ID
832272
Gene Name
DNA polymerase epsilon subunit B2
Gene Aliases
AT5G22110, ATDPB2, CYCLOPS 2, CYL2, DNA polymerase epsilon subunit B2
Chromosome Location
Chr.5: 7330825 - 7334282 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343719.1 NP_001332528.1 10 - 11 1318 143
NM_147892.4 NP_680197.2 10 - 11 1318 143
GenBank mRNA BT022108.1 - 10 - 11 1239 143

View Full Product Details