Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-A5
Entrez Gene ID
842851
Gene Name
phloem protein 2 A5
Gene Aliases
AT1G65390, AT1G65400, ATPP2-A5, T8F5.18, T8F5_18, phloem protein 2 A5
Chromosome Location
Chr.1: 24292372 - 24294740 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334203.1 NP_001319325.1 2 - 3 735 107
NM_001334204.1 NP_001322028.1 2 - 3 747 107
NM_105213.4 NP_176718.2 2 - 3 853 107
GenBank mRNA AF325110.1 - 2 - 3 659 107
AK176581.1 - 2 - 3 747 107

View Full Product Details