Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEG4
Entrez Gene ID
842875
Gene Name
DegP protease 4
Gene Aliases
AT1G65640, DegP protease 4, DegP4, F1E22.2, degradation of periplasmic proteins 4
Chromosome Location
Chr.1: 24410851 - 24413527 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334218.1 NP_001322516.1 5 - 6 1139 106
NM_105237.2 NP_564857.2 5 - 6 1139 106

View Full Product Details