Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ETR1
- Entrez Gene ID
-
842951
- Gene Name
- Signal transduction histidine kinase, hybrid-type, ethylene sensor
- Gene Aliases
-
AT1G66340, AtETR1, EIN1, ETHYLENE INSENSITIVE 1, ETHYLENE RESPONSE, ETHYLENE RESPONSE 1, ETR, HISTIDINE KINASE ETR1, T27F4.9, T27F4_9
- Chromosome Location
-
Chr.1: 24734162 - 24737756 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105305.4 |
NP_176808.3 |
3 - 4 |
1440 |
131 |
- Gene Symbol
- AT2G45180
- Entrez Gene ID
-
819126
- Gene Name
- Bifunctional inhibitor/lipid-transfer protein/seed storage 2S albumin superfamily protein
- Gene Aliases
-
AT2G45180, F4L23.31
- Chromosome Location
-
Chr.2: 18626188 - 18627157 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK230334.1 |
- |
- |
910 |
131 |
View Full Product Details