Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SWEETIE
Entrez Gene ID
843034
Gene Name
HEAT repeat-containing protein
Gene Aliases
AT1G67140, F5A8.12, F5A8_12
Chromosome Location
Chr.1: 25100848 - 25117531 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198408.1 NP_001185337.1 22 - 23 2534 79
NM_001334274.1 NP_001319331.1 22 - 23 2606 79
NM_105384.8 NP_176885.7 22 - 23 2642 79

View Full Product Details