Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASY1
Entrez Gene ID
843058
Gene Name
DNA-binding HORMA family protein
Gene Aliases
ASYNAPTIC 1, AT1G67370, ATASY1, F1N21.19
Chromosome Location
Chr.1: 25239117 - 25243837 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105405.1 NP_564896.1 21 - 22 1865 58

View Full Product Details