The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001334301.1 | NP_001319333.1 | 5 - 6 | 432 | 137 |
NM_001334302.1 | NP_001320390.1 | 5 - 6 | 483 | 137 | |
NM_001334303.1 | NP_001320389.1 | 5 - 6 | 464 | 137 | |
NM_001334304.1 | NP_001320393.1 | 5 - 6 | 460 | 137 | |
NM_001334305.1 | NP_001320391.1 | 5 - 6 | 459 | 137 | |
NM_001334306.1 | NP_001320392.1 | 5 - 6 | 473 | 137 | |
NM_105417.4 | NP_176917.2 | 5 - 6 | 432 | 137 | |
GenBank mRNA | AB114052.1 | - | 4 - 5 | 300 | 137 |