Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REV3
Entrez Gene ID
843071
Gene Name
recovery protein 3
Gene Aliases
AT1G67500, ATREV3, T1F15.3, T1F15_3, recovery protein 3
Chromosome Location
Chr.1: 25287356 - 25296988 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105417.4 NP_176917.2 6 - 7 549 68
GenBank mRNA AB114052.1 - 5 - 6 417 68

View Full Product Details