Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPR
Entrez Gene ID
843129
Gene Name
hydroxypyruvate reductase
Gene Aliases
AT1G68010, ATHPR1, hydroxypyruvate reductase
Chromosome Location
Chr.1: 25493193 - 25496053 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198420.1 NP_001185349.1 5 - 6 778 93
NM_001334339.1 NP_001320607.1 5 - 6 756 93
NM_105471.4 NP_176968.1 5 - 6 781 93
GenBank mRNA AF370221.1 - 5 - 6 621 93
AK226564.1 - - 598 93
AY099552.1 - 5 - 6 613 93
AY113871.1 - 5 - 6 556 93
BT001237.1 - 5 - 6 556 93

View Full Product Details