Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPR
Entrez Gene ID
843129
Gene Name
hydroxypyruvate reductase
Gene Aliases
AT1G68010, ATHPR1, hydroxypyruvate reductase
Chromosome Location
Chr.1: 25493193 - 25496053 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198420.1 NP_001185349.1 9 - 10 1126 64
NM_001334339.1 NP_001320607.1 9 - 10 1101 64
NM_105471.4 NP_176968.1 9 - 10 1126 64
GenBank mRNA AF370221.1 - 9 - 10 966 64
AK226564.1 - - 943 64
AY099552.1 - 9 - 10 958 64
AY113871.1 - 9 - 10 901 64
BT001237.1 - 9 - 10 901 64

View Full Product Details