Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAR1
Entrez Gene ID
843138
Gene Name
ZIP metal ion transporter family
Gene Aliases
AT1G68100, IAA-ALANINE RESISTANCE PROTEIN 1, IAA-ALANINE RESISTANT 1, T23K23.5, T23K23_5
Chromosome Location
Chr.1: 25521100 - 25524250 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334346.1 NP_001322211.1 4 - 5 719 110
NM_105480.3 NP_564921.1 4 - 5 728 110
GenBank mRNA AF216524.1 - 4 - 5 553 110
AK229148.1 - 4 - 5 610 110
AY084533.1 - 4 - 5 624 110
BT001920.1 - 4 - 5 503 110

View Full Product Details