Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENDO2
Entrez Gene ID
843158
Gene Name
endonuclease 2
Gene Aliases
AT1G68290, AtBFN2, T22E19.8, T22E19_8, endonuclease 2
Chromosome Location
Chr.1: 25596671 - 25598438 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105500.2 NP_176996.1 8 - 9 826 99

View Full Product Details