Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMT3
Entrez Gene ID
843313
Gene Name
chromomethylase 3
Gene Aliases
AT1G69770, T6C23.3, T6C23_3, chromomethylase 3
Chromosome Location
Chr.1: 26248118 - 26253687 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105645.4 NP_177135.1 11 - 12 1515 117

View Full Product Details