Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT1G71810
Entrez Gene ID
843511
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT1G71810, F14O23.19, F14O23_19
Chromosome Location
Chr.1: 27002414 - 27008184 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105839.3 NP_565025.1 10 - 11 1416 63
GenBank mRNA AY039896.1 - 10 - 11 1319 63
AY094006.1 - 10 - 11 1196 63

View Full Product Details