Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VND7
Entrez Gene ID
843524
Gene Name
vascular related NAC-domain protein 7
Gene Aliases
ANAC030, AT1G71930, Arabidopsis NAC domain containing protein 30, F17M19.8, F17M19_8, vascular related NAC-domain protein 7
Chromosome Location
Chr.1: 27074702 - 27077991 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334512.1 NP_001323397.1 2 - 3 512 145
NM_105851.2 NP_177338.1 1 - 2 345 145
GenBank mRNA AY085424.1 - 1 - 2 225 145
BT026447.1 - 1 - 2 160 145

View Full Product Details