Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIM23-2
Entrez Gene ID
843607
Gene Name
translocase inner membrane subunit 23-2
Gene Aliases
AT1G72750, ATTIM23-2, translocase inner membrane subunit 23-2
Chromosome Location
Chr.1: 27383320 - 27384524 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105934.2 NP_177419.1 1 - 1 965 152
GenBank mRNA AY042892.1 - 1 - 1 888 152
AY088456.1 - 1 - 1 903 152

Gene Symbol
AT5G41685
Entrez Gene ID
834171
Gene Name
Mitochondrial outer membrane translocase complex, subunit Tom7
Gene Aliases
AT5G41685
Chromosome Location
Chr.5: 16669149 - 16669723 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK230298.1 - - 154 152

View Full Product Details