Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl4
Entrez Gene ID
843665
Gene Name
serine carboxypeptidase-like 4
Gene Aliases
AT1G73310, T18K17.2, T18K17_2, serine carboxypeptidase-like 4
Chromosome Location
Chr.1: 27562441 - 27565177 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334584.1 NP_001322854.1 7 - 8 824 104
NM_001334585.1 NP_001322853.1 6 - 7 700 104
NM_105990.2 NP_177474.1 7 - 8 824 104

View Full Product Details