Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUS6
Entrez Gene ID
843672
Gene Name
sucrose synthase 6
Gene Aliases
ARABIDOPSIS THALIANA SUCROSE SYNTHASE 6, AT1G73370, ATSUS6, T9L24.42, T9L24_42, sucrose synthase 6
Chromosome Location
Chr.1: 27584364 - 27588978 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198461.1 NP_001185390.1 11 - 12 2526 115
NM_001334591.1 NP_001319374.1 12 - 13 2837 115
NM_105997.2 NP_177480.1 11 - 12 2773 115
GenBank mRNA KJ138940.1 - 11 - 12 2410 115

View Full Product Details