Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF8
Entrez Gene ID
833672
Gene Name
auxin response factor 8
Gene Aliases
AT5G37020, ATARF8, K15O15.1, K15O15_1, auxin response factor 8
Chromosome Location
Chr.5: 14629453 - 14634454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085203.1 NP_001078672.1 6 - 7 673 77
NM_001344162.1 NP_001331077.1 5 - 6 1201 77
NM_123060.3 NP_198518.1 6 - 7 1248 77
GenBank mRNA AF042196.1 - 6 - 7 673 77
AY669790.1 - 6 - 7 550 77

View Full Product Details