Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GORK
Entrez Gene ID
833728
Gene Name
gated outwardly-rectifying K+ channel
Gene Aliases
AT5G37500, MPA22.4, MPA22_4, gated outwardly-rectifying K+ channel
Chromosome Location
Chr.5: 14889245 - 14895233 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344207.1 NP_001332118.1 5 - 6 1464 86
NM_001344208.1 NP_001332117.1 4 - 5 1176 86
NM_001344209.1 NP_001332119.1 5 - 6 1464 86
NM_123109.5 NP_198566.2 5 - 6 1464 86

View Full Product Details