Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC
Entrez Gene ID
819579
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT3G04240, T6K12.14, T6K12_14, secret agent
Chromosome Location
Chr.3: 1113914 - 1120933 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337509.1 NP_001326045.1 4 - 5 603 74
NM_111295.4 NP_187074.1 10 - 11 1203 74
GenBank mRNA AF441079.1 - 10 - 11 1121 74
AK226884.1 - 10 - 11 1146 74
AY090938.1 - 10 - 11 1142 74
AY117340.1 - 10 - 11 992 74
KJ138841.1 - 10 - 11 992 74

View Full Product Details