Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZIFL2
Entrez Gene ID
823490
Gene Name
zinc induced facilitator-like 2
Gene Aliases
AT3G43790, zinc induced facilitator-like 2
Chromosome Location
Chr.3: 15655319 - 15660126 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339105.1 NP_001326438.1 8 - 9 744 101
NM_001339106.1 NP_001326437.1 8 - 9 740 101
NM_114247.4 NP_189965.2 8 - 9 744 101
NM_202653.1 NP_974382.1 8 - 9 679 101
NM_202654.1 NP_974383.1 8 - 9 679 101
GenBank mRNA BT030378.1 - 8 - 9 620 101

View Full Product Details