Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDA9
Entrez Gene ID
823594
Gene Name
histone deacetylase 9
Gene Aliases
AT3G44680, AtHDA9, HDA09, HISTONE DEACETYLASE, histone deacetylase 9
Chromosome Location
Chr.3: 16226519 - 16230009 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114336.4 NP_190054.2 7 - 8 971 90
GenBank mRNA BT002003.1 - 7 - 8 881 90
BT006576.1 - 7 - 8 714 90

View Full Product Details