Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLA2-ALPHA
Entrez Gene ID
815261
Gene Name
Phospholipase A2 family protein
Gene Aliases
AT2G06925, ATSPLA2-ALPHA, PHOSPHOLIPASE A2-ALPHA
Chromosome Location
Chr.2: 2842020 - 2843386 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335309.1 NP_001324649.1 1 - 2 222 69
NM_126670.3 NP_565337.1 1 - 2 270 69
GenBank mRNA AK221680.1 - 1 - 2 148 69
AY088532.1 - 1 - 2 185 69
AY136317.1 - 1 - 2 148 69
AY344842.1 - 1 - 2 173 69
BT002200.1 - 1 - 2 96 69

View Full Product Details