Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRP3B
Entrez Gene ID
815898
Gene Name
dynamin related protein
Gene Aliases
ADL2B, AT2G14120, T22C12.5, T22C12_5, dynamin related protein
Chromosome Location
Chr.2: 5953940 - 5960263 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161044.1 NP_001154516.1 19 - 20 2282 69
NM_001335415.1 NP_001323471.1 18 - 19 2224 69
NM_126984.4 NP_565362.1 18 - 19 2350 69
NM_126985.6 NP_565363.2 18 - 19 2195 69

View Full Product Details