Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRP3B
Entrez Gene ID
815898
Gene Name
dynamin related protein
Gene Aliases
ADL2B, AT2G14120, T22C12.5, T22C12_5, dynamin related protein
Chromosome Location
Chr.2: 5953940 - 5960263 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161044.1 NP_001154516.1 1 - 2 459 94
NM_001335415.1 NP_001323471.1 1 - 2 488 94
NM_126984.4 NP_565362.1 1 - 2 614 94
NM_126985.6 NP_565363.2 1 - 2 459 94

View Full Product Details