The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_100149.4 | NP_171769.1 | 8 - 9 | 1362 | 75 |
NM_202014.1 | NP_973743.1 | 8 - 9 | 1239 | 75 | |
GenBank mRNA | AY070068.1 | - | 8 - 9 | 1236 | 75 |
AY096437.1 | - | 8 - 9 | 1099 | 75 |