Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AXR1
Entrez Gene ID
839286
Gene Name
NAD(P)-binding Rossmann-fold superfamily protein
Gene Aliases
AT1G05180, AUXIN RESISTANT 1, AUXIN RESISTANT PROTEIN 1, YUP8H12.21, YUP8H12_21
Chromosome Location
Chr.1: 1498114 - 1501953 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035893.1 NP_001030970.1 8 - 9 814 125
NM_100396.4 NP_172010.1 7 - 8 1041 125
NM_202032.2 NP_973761.1 8 - 9 814 125
GenBank mRNA AY050379.1 - 7 - 8 901 125
L13922.1 - 7 - 8 880 125

View Full Product Details