Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY39
Entrez Gene ID
819625
Gene Name
WRKY DNA-binding protein 39
Gene Aliases
AT3G04670, ATWRKY39, F7O18.30, F7O18_30, WRKY DNA-BINDING PROTEIN 39, WRKY DNA-binding protein 39
Chromosome Location
Chr.3: 1266056 - 1268378 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035557.2 NP_001030634.1 1 - 2 1384 64
NM_111339.3 NP_566236.1 1 - 2 1412 64
GenBank mRNA AF404860.1 - 1 - 2 725 64
AK316972.1 - 1 - 2 1251 64
AY053420.1 - 1 - 2 1321 64
AY133586.1 - 1 - 2 725 64

View Full Product Details