Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEK2
Entrez Gene ID
819641
Gene Name
NIMA-related kinase 2
Gene Aliases
AT3G04810, ATNEK2, NIMA-RELATED KINASE2, NIMA-related kinase 2, T9J14.24, T9J14_24
Chromosome Location
Chr.3: 1317245 - 1321294 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337542.1 NP_001327439.1 10 - 11 1238 99
NM_111353.4 NP_187132.1 11 - 12 886 99
NM_202492.3 NP_974221.2 10 - 11 878 99
GenBank mRNA BT000875.1 - 9 - 10 564 99

View Full Product Details