Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD9
Entrez Gene ID
819713
Gene Name
cell cycle checkpoint control protein family
Gene Aliases
AT3G05480, ATRAD9, F22F7.7, F22F7_7
Chromosome Location
Chr.3: 1585272 - 1589292 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202886.1 NP_001189815.1 5 - 6 723 104
NM_001337585.1 NP_001319478.1 4 - 5 702 104
NM_001337586.1 NP_001319479.1 4 - 5 685 104
GenBank mRNA BT029213.1 - 4 - 5 591 104

View Full Product Details