Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IBR3
Entrez Gene ID
819865
Gene Name
acyl-CoA dehydrogenase-like protein
Gene Aliases
AT3G06800, AT3G06810, IBA-RESPONSE 3
Chromosome Location
Chr.3: 2146243 - 2150851 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111561.5 NP_187337.2 3 - 4 699 114
GenBank mRNA AY091014.1 - 3 - 4 462 114
BT002365.1 - 3 - 4 408 114

View Full Product Details