Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFP2
Entrez Gene ID
819870
Gene Name
multifunctional protein 2
Gene Aliases
AT3G06860, ATMFP2, F3E22.20, MULTIFUNCTIONAL PROTEIN 2, multifunctional protein 2
Chromosome Location
Chr.3: 2161735 - 2166357 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111566.4 NP_187342.1 8 - 9 1001 74
GenBank mRNA AF123254.1 - 8 - 9 810 74
AY062621.1 - 8 - 9 867 74

View Full Product Details